Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028721.3 | NP_082997.3 | 25 - 26 | 3922 | 103 | |
GenBank mRNA | AK014393.1 | - | 10 - 11 | 1482 | 103 | |
AK173327.1 | - | 25 - 26 | 3501 | 103 | ||
AY259499.1 | - | 25 - 26 | 3859 | 103 | ||
BC115724.1 | - | 25 - 26 | 3449 | 40086844 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |