Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028721.3 | NP_082997.3 | 22 - 23 | 3423 | 127 | |
GenBank mRNA | AK014393.1 | - | 7 - 8 | 983 | 127 | |
AK133217.1 | - | 22 - 23 | 3424 | 127 | ||
AK173327.1 | - | 22 - 23 | 3002 | 127 | ||
AY259499.1 | - | 22 - 23 | 3360 | 127 | ||
BC115724.1 | - | 22 - 23 | 2950 | 40086844 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |