Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007567.2 | NP_031593.2 | 10 - 11 | 11970 | 121 | |
GenBank mRNA | AK010775.1 | - | 3 - 4 | 265 | 121 | |
AK129141.1 | - | 6 - 7 | 3530 | 121 | ||
AK135719.1 | - | 3 - 4 | 258 | 121 | ||
AK160649.1 | - | 3 - 4 | 292 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |