Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007567.2 | NP_031593.2 | 9 - 10 | 11915 | 124 | |
GenBank mRNA | AK010775.1 | - | 2 - 3 | 210 | 124 | |
AK129141.1 | - | 5 - 6 | 3475 | 124 | ||
AK135719.1 | - | 2 - 3 | 203 | 124 | ||
AK160649.1 | - | 2 - 3 | 237 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |