Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008243.3 | NP_032269.3 | 1 - 2 | 92 | 69 | |
GenBank mRNA | AK040934.1 | - | 1 - 2 | 76 | 69 | |
AK134222.1 | - | 1 - 2 | 75 | 69 | ||
AK144824.1 | - | 1 - 2 | 55 | 69 | ||
BC010551.1 | - | 1 - 2 | 77 | 4195560 | 69 | |
BC051393.1 | - | 1 - 2 | 61 | 5055055 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |