Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025692.3 | NP_079968.2 | 6 - 7 | 746 | 69 | |
GenBank mRNA | AK077825.1 | - | 6 - 7 | 727 | 69 | |
AK151823.1 | - | 6 - 7 | 703 | 69 | ||
AK152029.1 | - | 6 - 7 | 741 | 69 | ||
AK168907.1 | - | 6 - 7 | 740 | 69 | ||
AK207610.1 | - | 4 - 5 | 310 | 69 | ||
BC019764.1 | - | 6 - 7 | 623 | 3989119 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |