Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133977.2 | NP_598738.1 | 7 - 8 | 936 | 104 | |
GenBank mRNA | AF440692.1 | - | 6 - 7 | 866 | 104 | |
AK085754.1 | - | 7 - 8 | 921 | 104 | ||
AK142599.1 | - | 926 | 104 | |||
AK146549.1 | - | 7 - 8 | 921 | 104 | ||
AK149559.1 | - | 7 - 8 | 921 | 104 | ||
AK149595.1 | - | 7 - 8 | 921 | 104 | ||
AK150782.1 | - | 7 - 8 | 969 | 104 | ||
AK168368.1 | - | 7 - 8 | 923 | 104 | ||
AK168405.1 | - | 7 - 8 | 923 | 104 | ||
AK168419.1 | - | 7 - 8 | 921 | 104 | ||
AK168938.1 | - | 7 - 8 | 921 | 104 | ||
AK169187.1 | - | 7 - 8 | 920 | 104 | ||
BC008559.1 | - | 5 - 6 | 658 | 104 | ||
BC012313.1 | - | 7 - 8 | 936 | 3981648 | 104 | |
BC018573.1 | - | 2 - 3 | 206 | 104 | ||
BC020295.1 | - | 7 - 8 | 905 | 104 | ||
BC022986.1 | - | 7 - 8 | 914 | 5123741 | 104 | |
BC058216.1 | - | 7 - 8 | 902 | 104 | ||
BC058218.1 | - | 7 - 8 | 915 | 104 | ||
BC092046.1 | - | 7 - 8 | 897 | 3594705 | 104 | |
S67217.1 | - | 1 - 2 | 67 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |