Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148930.3 | NP_683732.1 | 23 - 24 | 2361 | 69 | |
GenBank mRNA | AJ309168.1 | - | 22 - 23 | 2196 | 69 | |
AK146880.1 | - | 23 - 24 | 2356 | 69 | ||
BC003988.1 | - | 13 - 14 | 1310 | 69 | ||
BC005719.1 | - | 18 - 19 | 1966 | 69 | ||
BC023854.1 | - | 23 - 24 | 2346 | 5346780 | 69 | |
BC031899.1 | - | 23 - 24 | 2346 | 5362121 | 69 | |
BC042419.1 | - | 21 - 22 | 5597 | 69 | ||
BC043058.1 | - | 23 - 24 | 2298 | 6419695 | 69 | |
BC054729.1 | - | 23 - 24 | 2349 | 6419758 | 69 | |
BC054820.1 | - | 21 - 22 | 5075 | 69 | ||
BC060985.1 | - | 22 - 23 | 2265 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |