Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146226.2 | NP_666338.2 | 12 - 13 | 1191 | 68 | |
GenBank mRNA | AK041271.1 | - | 9 - 10 | 1831 | 68 | |
AK149671.1 | - | 12 - 13 | 1195 | 68 | ||
AK149713.1 | - | 12 - 13 | 1193 | 68 | ||
AK218199.1 | - | 2 - 3 | 168 | 68 | ||
BC005716.1 | - | 11 - 12 | 1317 | 68 | ||
BC023937.1 | - | 2 - 3 | 160 | 68 | ||
BC025494.1 | - | 12 - 13 | 1173 | 5310043 | 68 | |
BC026594.1 | - | 7 - 8 | 724 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |