Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098237.1 | NP_001091707.1 | 1 - 2 | 160 | 67 | |
GenBank mRNA | BC012410.1 | - | 1 - 2 | 135 | 67 | |
CF983027.1 | - | 1 - 2 | 160 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |