Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190401.2 | NP_001177330.2 | 13 - 14 | 1620 | 111 | |
GenBank mRNA | AK148805.1 | - | 11 - 12 | 1066 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |