Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039657.2 | NP_001034746.1 | 1 - 2 | 477 | 70 | |
NM_001039658.2 | NP_001034747.1 | 1 - 2 | 477 | 70 | ||
GenBank mRNA | AB057423.1 | - | 1 - 2 | 465 | 70 | |
AK015724.1 | - | 1 - 2 | 418 | 70 | ||
AK015732.1 | - | 1 - 2 | 450 | 70 | ||
AK161511.1 | - | 1 - 2 | 478 | 70 | ||
BC024377.1 | - | 1 - 2 | 453 | 4976492 | 70 | |
U77383.1 | - | 1 - 2 | 114 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |