Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271496.1 | NP_001258425.1 | 10 - 11 | 1522 | 85 | |
NM_013490.4 | NP_038518.2 | 9 - 10 | 1468 | 85 | ||
NR_073190.1 | 11 - 12 | 1582 | 85 | |||
GenBank mRNA | AB011002.1 | - | 9 - 10 | 1327 | 85 | |
AK005057.1 | - | 11 - 12 | 1583 | 85 | ||
AK166465.1 | - | 9 - 10 | 1409 | 85 | ||
AK184298.1 | - | 3 - 4 | 262 | 85 | ||
BC004719.1 | - | 7 - 8 | 658 | 85 | ||
BC060218.1 | - | 8 - 9 | 3548 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |