Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073190.1 | 3 - 4 | 813 | 75 | ||
GenBank mRNA | AK005057.1 | - | 3 - 4 | 814 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |