Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026347.3 | NP_080623.2 | 3 | 234 | 93 | |
GenBank mRNA | AK005287.1 | - | 3 | 235 | 93 | |
AK011299.1 | - | 3 | 235 | 93 | ||
AK014743.1 | - | 3 | 212 | 93 | ||
BC060949.1 | - | 3 | 237 | 6533986 | 93 | |
BC087901.1 | - | 3 | 205 | 30303437 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |