Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011237.2 | NP_035367.1 | 4 - 5 | 439 | 97 | |
GenBank mRNA | AF045663.1 | - | 4 - 5 | 439 | 97 | |
AK134293.1 | - | 4 - 5 | 452 | 97 | ||
AK138973.1 | - | 4 - 5 | 444 | 97 | ||
AK165833.1 | - | 4 - 5 | 474 | 97 | ||
BC082556.1 | - | 4 - 5 | 378 | 30538342 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |