Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081278.2 | NP_001074747.2 | 2 - 3 | 1838 | 64 | |
GenBank mRNA | AK031120.1 | - | 2 - 3 | 1682 | 64 | |
AK039337.1 | - | 2 - 3 | 1839 | 64 | ||
AK044719.1 | - | 2 - 3 | 1480 | 64 | ||
AK220347.1 | - | 1 - 2 | 245 | 64 | ||
BC064433.1 | - | 2 - 3 | 1462 | 64 | ||
BC080762.1 | - | 2 - 3 | 1456 | 64 | ||
BC117869.1 | - | 2 - 3 | 1314 | 40095526 | 64 | |
BC117870.1 | - | 2 - 3 | 1314 | 40095531 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |