Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194334.2 | NP_919315.2 | 2 - 3 | 577 | 143 | |
GenBank mRNA | AK089175.1 | - | 2 - 3 | 542 | 143 | |
AK129273.1 | - | 2 - 3 | 563 | 143 | ||
AK148382.1 | - | 2 - 3 | 580 | 143 | ||
AK156788.1 | - | 2 - 3 | 564 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |