Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 1 - 2 | 919 | 66 | |
GenBank mRNA | AF237953.1 | - | 1 - 2 | 160 | 66 | |
AK171516.1 | - | 1 - 2 | 706 | 66 | ||
BC054751.1 | - | 1 - 2 | 198 | 66 | ||
BC055289.1 | - | 2 - 3 | 722 | 66 | ||
BC096538.1 | - | 1 - 2 | 919 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |