Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 23 - 24 | 4574 | 69 | |
GenBank mRNA | AF237953.1 | - | 21 - 22 | 2591 | 69 | |
AK220471.1 | - | 1 - 2 | 472 | 69 | ||
BC025838.1 | - | 17 - 18 | 2077 | 69 | ||
BC038056.1 | - | 18 - 19 | 2682 | 69 | ||
BC054751.1 | - | 23 - 24 | 3853 | 69 | ||
BC055289.1 | - | 24 - 25 | 4377 | 69 | ||
BC096538.1 | - | 23 - 24 | 4574 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |