Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164352.1 | NP_001157824.1 | 5 - 6 | 939 | 62 | |
NM_021474.3 | NP_067449.3 | 6 - 7 | 698 | 62 | ||
GenBank mRNA | AF104223.1 | - | 5 - 6 | 653 | 62 | |
AF109122.1 | - | 5 - 6 | 912 | 62 | ||
AK075585.1 | - | 6 - 7 | 687 | 62 | ||
BC012269.1 | - | 5 - 6 | 836 | 3980048 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |