Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024560.2 | NP_001019731.1 | 2 - 3 | 265 | 69 | |
GenBank mRNA | AK037118.1 | - | 272 | 69 | ||
AK047234.1 | - | 2 - 3 | 240 | 69 | ||
AK143441.1 | - | 2 - 3 | 283 | 69 | ||
BC048922.1 | - | 2 - 3 | 265 | 6311864 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |