Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024560.2 | NP_001019731.1 | 4 - 5 | 493 | 78 | |
GenBank mRNA | AK037118.1 | - | 484 | 78 | ||
AK047234.1 | - | 4 - 5 | 468 | 78 | ||
AK143441.1 | - | 3 - 4 | 605 | 78 | ||
BC048922.1 | - | 4 - 5 | 493 | 6311864 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |