Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024560.2 | NP_001019731.1 | 9 - 10 | 942 | 100 | |
GenBank mRNA | AK047234.1 | - | 9 - 10 | 918 | 100 | |
BC048922.1 | - | 9 - 10 | 942 | 6311864 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |