Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF191551.1 | - | 2 - 3 | 393 | 88 | |
BC144726.1 | - | 2 - 3 | 831 | 9053274 | 88 | |
U95167.1 | - | 2 - 3 | 845 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |