Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114977.1 | NP_001108449.1 | 19 - 20 | 2078 | 111 | |
NM_026476.2 | NP_080752.2 | 18 - 19 | 1946 | 111 | ||
GenBank mRNA | AK029898.1 | - | 19 - 20 | 1978 | 111 | |
BC031497.1 | - | 19 - 20 | 1938 | 111 | ||
BC068265.1 | - | 18 - 19 | 1962 | 30022109 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |