Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114595.1 | NP_001108067.1 | 14 - 15 | 2193 | 90 | |
NM_001114596.1 | NP_001108068.1 | 15 - 16 | 2229 | 90 | ||
NM_001114597.1 | NP_001108069.1 | 15 - 16 | 5061 | 90 | ||
NM_053252.3 | NP_444482.2 | 15 - 16 | 2538 | 90 | ||
GenBank mRNA | AF305087.1 | - | 15 - 16 | 5008 | 90 | |
AF305088.1 | - | 15 - 16 | 2482 | 90 | ||
AF305089.1 | - | 14 - 15 | 2137 | 90 | ||
AF305090.1 | - | 15 - 16 | 2173 | 90 | ||
AK087817.1 | - | 11 - 12 | 1780 | 90 | ||
BC004660.1 | - | 15 - 16 | 2478 | 3497913 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |