Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002896.3 | 7 - 8 | 310 | 99 | ||
GenBank mRNA | AK043166.1 | - | 7 - 8 | 311 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |