Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021567.5 | NP_067542.3 | 6 - 7 | 618 | 93 | |
GenBank mRNA | AF176328.1 | - | 6 - 7 | 504 | 93 | |
AK004580.1 | - | 6 - 7 | 569 | 93 | ||
AK005383.1 | - | 6 - 7 | 567 | 93 | ||
AK077632.1 | - | 6 - 7 | 568 | 93 | ||
AK082005.1 | - | 6 - 7 | 568 | 93 | ||
AK178757.1 | - | 2 - 3 | 237 | 93 | ||
AK179721.1 | - | 2 - 3 | 238 | 93 | ||
AK187694.1 | - | 2 - 3 | 237 | 93 | ||
AK188206.1 | - | 2 - 3 | 238 | 93 | ||
AK188263.1 | - | 2 - 3 | 237 | 93 | ||
AK189619.1 | - | 2 - 3 | 237 | 93 | ||
BC010694.1 | - | 6 - 7 | 507 | 2654291 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |