Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900025892.mm.1 |
NM_172774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025892.mm.1 |
NM_172774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025892.mm.1 |
TC0900003073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025893.mm.1 |
NM_172774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025893.mm.1 |
NM_172774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025893.mm.1 |
TC0900003073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025890.mm.1 |
NM_172774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025890.mm.1 |
NM_172774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025890.mm.1 |
TC0900003073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025887.mm.1 |
NM_172774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025887.mm.1 |
NM_172774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025887.mm.1 |
TC0900003073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025891.mm.1 |
NM_172774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025891.mm.1 |
NM_172774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025891.mm.1 |
TC0900003073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025888.mm.1 |
NM_172774.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025888.mm.1 |
NM_172774.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025888.mm.1 |
TC0900003073.mm.1 |
MTA TC |