Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080129.2 | NP_001073598.1 | 3 - 4 | 866 | 84 | |
NM_001080130.2 | NP_001073599.1 | 3 - 4 | 866 | 84 | ||
NM_001080131.2 | NP_001073600.1 | 3 - 4 | 866 | 84 | ||
NM_001080132.2 | NP_001073601.1 | 3 - 4 | 866 | 84 | ||
NM_001283048.1 | NP_001269977.1 | 3 - 4 | 549 | 84 | ||
GenBank mRNA | AA041637.1 | - | 1 - 2 | 93 | 84 | |
AK011988.1 | - | 3 - 4 | 359 | 84 | ||
AK011992.1 | - | 3 - 4 | 359 | 84 | ||
AK141845.1 | - | 3 - 4 | 816 | 84 | ||
AK146903.1 | - | 3 - 4 | 833 | 84 | ||
AK150305.1 | - | 3 - 4 | 814 | 84 | ||
AK150515.1 | - | 3 - 4 | 801 | 84 | ||
AK151936.1 | - | 3 - 4 | 818 | 84 | ||
AK152737.1 | - | 3 - 4 | 818 | 84 | ||
AK153281.1 | - | 3 - 4 | 801 | 84 | ||
AK159161.1 | - | 3 - 4 | 462 | 84 | ||
AK165282.1 | - | 3 - 4 | 816 | 84 | ||
AK165851.1 | - | 3 - 4 | 823 | 84 | ||
AK192607.1 | - | 3 - 4 | 305 | 84 | ||
BE852115.1 | - | 3 - 4 | 307 | 84 | ||
CN668856.1 | - | 1 - 2 | 69 | 84 | ||
U39073.1 | - | 3 - 4 | 769 | 84 | ||
U39074.1 | - | 3 - 4 | 797 | 84 | ||
U39075.1 | - | 3 - 4 | 797 | 84 | ||
U39076.1 | - | 3 - 4 | 797 | 84 | ||
U39077.1 | - | 3 - 4 | 797 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |