Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967153 |
AK136565.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
CD349605.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
AK029959.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
AK014958.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
AK029878.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
NM_029858.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
CD349605.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
AK029959.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
AK014958.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
AK029878.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P208435 |
NM_029858.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
843775 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
411309 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
248866 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371179 |
XM_205565.5 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371179 |
XM_918262.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
741329 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
111977 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
82315 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
840383 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
167170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
785858 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
509365 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879641 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729335 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
526484 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
661254 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
433959 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153630 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
94902 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402802 |
AK078933.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402802 |
BC118519.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402802 |
L24191.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402802 |
NM_008118.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
282562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277398 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
53174 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66559 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P113514 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
CD349605.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
AK029959.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
AK014958.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
AK029878.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
NM_029858.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
CD349605.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
AK029959.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
AK014958.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
AK029878.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P208435 |
NM_029858.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967153 |
AK136565.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
CD349605.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
AK029959.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
AK014958.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
AK029878.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
NM_029858.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
CD349605.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
AK029959.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
AK014958.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
AK029878.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P208435 |
NM_029858.2 |
mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31428963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31429029 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428851 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428914 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428858 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428906 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428994 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428844 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429018 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429023 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428843 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428862 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428997 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428967 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428846 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13491663 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428887 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428852 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428854 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428874 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428973 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428889 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428872 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428867 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428892 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429007 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13270804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13493309 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13202298 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429025 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428866 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428920 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429013 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428897 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428917 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13189392 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428875 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428995 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429004 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428891 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428963 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428909 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429000 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13555379 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428900 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428970 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428868 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429042 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428908 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428893 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429011 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428883 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13440468 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428859 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428990 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428856 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429002 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13310219 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429015 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428999 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428863 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428916 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13202301 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13440459 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428989 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428904 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428996 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428912 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428886 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429039 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428988 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428965 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428878 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428884 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428885 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428848 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428968 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31429017 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428915 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31428969 |
- |
- |