Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286518.1 | NP_001273447.1 | 6 - 7 | 792 | 137 | |
GenBank mRNA | BC028439.1 | - | 5 - 6 | 657 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |