Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152822.3 | NP_690035.2 | 5 - 6 | 880 | 85 | |
GenBank mRNA | AK018059.1 | - | 5 - 6 | 810 | 85 | |
BC004735.1 | - | 4 - 5 | 472 | 85 | ||
BC032218.1 | - | 5 - 6 | 803 | 85 | ||
BC141155.1 | - | 5 - 6 | 817 | 9055767 | 85 | |
BQ573371.1 | - | 4 - 5 | 460 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |