Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252466.1 | NP_001239395.1 | 11 - 12 | 1139 | 99 | |
NM_001252467.1 | NP_001239396.1 | 8 - 9 | 779 | 99 | ||
NM_027933.3 | NP_082209.1 | 10 - 11 | 938 | 99 | ||
NR_045519.1 | 11 - 12 | 1048 | 99 | |||
GenBank mRNA | AK011541.1 | - | 3 - 4 | 299 | 99 | |
AK040481.1 | - | 12 - 13 | 1122 | 99 | ||
AK133949.1 | - | 10 - 11 | 785 | 99 | ||
AK155301.1 | - | 10 - 11 | 807 | 99 | ||
BC066005.1 | - | 10 - 11 | 779 | 99 | ||
BC145892.1 | - | 10 - 11 | 802 | 40131195 | 99 | |
BC145894.1 | - | 10 - 11 | 802 | 40131197 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |