Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152817.4 | NP_690030.3 | 11 - 12 | 1506 | 99 | |
GenBank mRNA | AK012131.1 | - | 11 - 12 | 1506 | 99 | |
AK030942.1 | - | 11 - 12 | 1507 | 99 | ||
AK049324.1 | - | 11 - 12 | 1507 | 99 | ||
AK168138.1 | - | 11 - 12 | 99 | |||
BC021912.1 | - | 10 - 11 | 1241 | 4503335 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |