Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152817.4 | NP_690030.3 | 8 - 9 | 1222 | 79 | |
GenBank mRNA | AK012131.1 | - | 8 - 9 | 1222 | 79 | |
AK049324.1 | - | 8 - 9 | 1223 | 79 | ||
AK168138.1 | - | 8 - 9 | 1213 | 79 | ||
BC021912.1 | - | 7 - 8 | 957 | 4503335 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |