Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P318577 |
NM_001163625.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P318577 |
AK010729.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P318577 |
NM_001163625.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P318577 |
AK010729.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
448235 |
AK047018.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
794291 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
AF073309.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
AK016561.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
AK171278.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
BC051962.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
BC086618.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
BC132303.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
365297 |
NM_018769.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
95992 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490084 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851913 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
123629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
207401 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347813 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837749 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894581 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381751 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
44414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
269195 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
595062 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
58794 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
109989 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84704 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155570 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P318577 |
NM_001163625.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P318577 |
AK010729.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P318577 |
NM_001163625.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P318577 |
AK010729.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P318577 |
NM_001163625.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P318577 |
AK010729.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P318577 |
NM_001163625.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P318577 |
AK010729.1 |
mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374288 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374299 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374293 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374262 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374290 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374274 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374306 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13243089 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374309 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374272 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374280 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374265 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374268 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374273 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13271181 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13167664 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374282 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374296 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374275 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374269 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374279 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374264 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13055375 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374304 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374266 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374286 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374326 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374295 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374323 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374302 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374291 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374310 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374303 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374305 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374292 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374307 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374277 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31374301 |
- |
- |