Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288586.1 | NP_001275515.1 | 5 - 6 | 554 | 113 | |
NM_010786.4 | NP_034916.1 | 6 - 7 | 639 | 113 | ||
GenBank mRNA | AK004719.1 | - | 6 - 7 | 598 | 113 | |
AK004881.1 | - | 6 - 7 | 566 | 113 | ||
AK088638.1 | - | 6 - 7 | 569 | 113 | ||
AK150530.1 | - | 5 - 6 | 427 | 113 | ||
AK152685.1 | - | 5 - 6 | 427 | 113 | ||
AK160069.1 | - | 5 - 6 | 430 | 113 | ||
AK168690.1 | - | 6 - 7 | 639 | 113 | ||
BC050902.1 | - | 6 - 7 | 555 | 6415937 | 113 | |
BC092270.1 | - | 6 - 7 | 592 | 4018419 | 113 | |
CN703560.1 | - | 5 - 6 | 496 | 113 | ||
U47934.1 | - | 4 - 5 | 345 | 113 | ||
X58876.1 | - | 6 - 7 | 546 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |