Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033367.3 | NP_001028539.1 | 7 - 8 | 2829 | 73 | |
GenBank mRNA | AK035300.1 | - | 4 - 5 | 2230 | 73 | |
AK143864.1 | - | 7 - 8 | 2830 | 73 | ||
AK171624.1 | - | 7 - 8 | 2824 | 73 | ||
BC144862.1 | - | 5 - 6 | 2586 | 73 | ||
BC145617.1 | - | 6 - 7 | 2623 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |