Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139301.2 | NP_647462.1 | 7 - 8 | 1713 | 107 | |
GenBank mRNA | AF407332.1 | - | 7 - 8 | 1713 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |