Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026410.3 | NP_080686.1 | 1 - 2 | 111 | 76 | |
GenBank mRNA | AK010540.1 | - | 1 - 2 | 78 | 76 | |
AK011701.1 | - | 1 - 2 | 111 | 76 | ||
BC052904.1 | - | 1 - 2 | 109 | 30065848 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |