Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026410.3 | NP_080686.1 | 2 - 3 | 205 | 72 | |
GenBank mRNA | AK010540.1 | - | 2 - 3 | 172 | 72 | |
AK011701.1 | - | 2 - 3 | 205 | 72 | ||
AK016305.1 | - | 2 - 3 | 319 | 72 | ||
BC052904.1 | - | 2 - 3 | 203 | 30065848 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |