Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030109.2 | NP_084385.2 | 3 - 4 | 320 | 60 | |
GenBank mRNA | AK011613.1 | - | 3 - 4 | 279 | 60 | |
AK012009.1 | - | 3 - 4 | 274 | 60 | ||
AK012643.1 | - | 3 - 4 | 289 | 60 | ||
AK019282.1 | - | 3 - 4 | 269 | 60 | ||
AK142136.1 | - | 3 - 4 | 290 | 60 | ||
AK146063.1 | - | 3 - 4 | 290 | 60 | ||
AK146540.1 | - | 3 - 4 | 285 | 60 | ||
AK146864.1 | - | 3 - 4 | 288 | 60 | ||
AK147687.1 | - | 3 - 4 | 288 | 60 | ||
AK149776.1 | - | 3 - 4 | 288 | 60 | ||
AK150961.1 | - | 3 - 4 | 277 | 60 | ||
AK151354.1 | - | 3 - 4 | 267 | 60 | ||
AK152327.1 | - | 3 - 4 | 285 | 60 | ||
AK152965.1 | - | 3 - 4 | 275 | 60 | ||
AK159893.1 | - | 3 - 4 | 277 | 60 | ||
AK211303.1 | - | 1 - 2 | 27 | 60 | ||
BC029745.1 | - | 3 - 4 | 255 | 60 | ||
BC045207.1 | - | 3 - 4 | 289 | 60 | ||
BC049118.1 | - | 3 - 4 | 313 | 6812422 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |