Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_001168655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_001168655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_001168656.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_001168656.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_001168657.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_001168657.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_007706.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
NM_007706.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020941.mm.1 |
TC1000002725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_001168655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_001168655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_001168656.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_001168656.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_001168657.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_001168657.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_007706.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
NM_007706.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020935.mm.1 |
TC1000002725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_001168655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_001168655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_001168656.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_001168656.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_001168657.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_001168657.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_007706.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
NM_007706.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020937.mm.1 |
TC1000002725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000020944.mm.1 |
NM_001168656.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020944.mm.1 |
NM_001168656.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000020944.mm.1 |
TC1000002725.mm.1 |
MTA TC |