Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013391.2 | NP_001013409.1 | 2 - 3 | 346 | 80 | |
GenBank mRNA | AK046856.1 | - | 2 - 3 | 80 | ||
AK048615.1 | - | 2 - 3 | 280 | 80 | ||
AK049749.1 | - | 2 - 3 | 344 | 80 | ||
AK077684.1 | - | 2 - 3 | 347 | 80 | ||
AK168764.1 | - | 2 - 3 | 311 | 80 | ||
BC068133.1 | - | 2 - 3 | 322 | 6813785 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |