Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010253.3 | NP_034383.1 | 1 - 2 | 146 | 62 | |
GenBank mRNA | AK133522.1 | - | 1 - 2 | 162 | 62 | |
BC044055.1 | - | 1 - 2 | 146 | 6476459 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |