Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177595.4 | NP_808263.2 | 4 - 5 | 746 | 88 | |
GenBank mRNA | AK034675.1 | - | 4 - 5 | 724 | 88 | |
BC137728.1 | - | 4 - 5 | 681 | 8860747 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |