Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153597.2 | NP_705825.2 | 4 - 5 | 827 | 72 | |
NR_027351.1 | 3 - 4 | 657 | 72 | |||
GenBank mRNA | AK038830.1 | - | 3 - 4 | 659 | 72 | |
AK048558.1 | - | 976 | 72 | |||
AK090183.1 | - | 3 - 4 | 1958 | 72 | ||
AK141582.1 | - | 614 | 72 | |||
BC027575.1 | - | 4 - 5 | 625 | 1364225 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |