Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181579.1 | NP_853557.1 | 10 - 11 | 1161 | 127 | |
GenBank mRNA | AF309775.1 | - | 9 - 10 | 1054 | 127 | |
AK036922.1 | - | 10 - 11 | 1166 | 128 | ||
AK086511.1 | - | 10 - 11 | 1162 | 127 | ||
AK137653.1 | - | 10 - 11 | 1160 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |